missing translation for 'onlineSavingsMsg'
Learn More

DL-1,2-Hexanediol, 98%

Product Code. 10646544 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10646544 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10646544 Supplier Thermo Scientific Acros Supplier No. 200132500
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6920-22-5 | C6H14O2 | 118.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6920-22-5
Melting Point -25.0°C
Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 223.0°C to 224.0°C
Flash Point 122°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H14O2
Linear Formula CH3(CH2)3CH(OH)CH2OH
Refractive Index 1.4410 to 1.4430
MDL Number MFCD00010737
Quantity 250 g
Beilstein 01, I, 251
Specific Gravity 0.95
Synonym 1,2-hexanediol, 1,2-dihydroxyhexane, dl-1,2-hexanediol, 1,2-hexane diol, dl-hexane-1,2-diol, 5,6-dihydroxyhexane, 1,2-hexylene glycol, acmc-209o6j, 2,2,6,6-tetrakis hydroxymet cyclohexanol
Solubility Information Solubility in water: soluble.
InChI Key FHKSXSQHXQEMOK-LURJTMIESA-N
SMILES CCCC[C@H](O)CO
IUPAC Name hexane-1,2-diol
Molecular Weight (g/mol) 118.18
PubChem CID 94335
Viscosity 87 mPa.s (20°C)
Formula Weight 118.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material DL-1, 2-Hexanediol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.