missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-1,4-Dithiothreitol, 99%, for biochemistry

Product Code. 10215550 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
Unit Size:
100g
1g
25g
500g
5g
This item is not returnable. View return policy

Product Code. 10215550

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,4-Dithiothreitol, CAS # 3483-12-3, is a reducing agent commonly used to reduce disulphide bonds in protein studies, such as western blot SDS-page electrophoresis. | CAS: 3483-12-03 | C4H10O2S2 | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • 1,4-Dithiothreitol (DTT or Cleland's reagent) reduces disulfide bonds in proteins, leading to loss of native conformation
  • DTT is not stable in solution and should be prepared in small aliquots prior to use; the use of EDTA in the solution can increase the half-life of DTT
  • Typically used at a final concentration of 0.1M for protein stabilization; recommended to create a 1M stock

Applications

  • Protects thiol groups from oxidation and acts as a chemical chelator in solution; maintains proteins in the denatured state
  • Common applications include SDS-page electrophoresis and protein isolation
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12-3-3483
MDL Number MFCD00004877
InChI Key VHJLVAABSRFDPM-IMJSIDKUSA-N
Synonym dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
PubChem CID 446094
ChEBI CHEBI:42170
IUPAC Name (2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
SMILES C(C(C(CS)O)O)S

Specifications

Absorbance (1 M soln. in water), (1 M soln. in water) at 0nm, 0.035 max. at 500nm
Melting Point 39°C to 44°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 0.99
Molecular Formula C4H10O2S2
Linear Formula HSCH2CH(OH)CH(OH)CH2SH
Quantity 25 g
Beilstein 01, III, 2360
Merck Index 15, 3419
Transmittance 99% min. (c=5, 660nm, water) 1cm cell
Molecular Weight (g/mol) 154.24
Formula Weight 154.24
Percent Purity ≥98.5%
Grade Biochemical
Physical Form Powder
Chemical Name or Material DL-1, 4-Dithiothreitol
Show More Show Less
compliance-icons
Product Identifier
  • DL-1,4-Dithiothreitol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.