missing translation for 'onlineSavingsMsg'
Learn More

DL-10-Camphorsulfonic acid, 98%

Product Code. 10780975 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10780975 2.5 kg Plastic bottle 2.5kg
10742041 100 g Plastic bottle 100g
10032550 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10780975 Supplier Thermo Scientific Acros Supplier No. 189070025
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5872-08-2 | C10H16O4S | 232.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8-2-5872
Molecular Formula C10H16O4S
Molecular Weight (g/mol) 232.29
MDL Number MFCD00074827
InChI Key MIOPJNTWMNEORI-UHFFFAOYNA-N
Synonym reychler's acid, camphorsulfonic acid, d-camphorsulfonic acid, camphersulfosaeure, --10-camphorsulfonic acid, 2-oxobornane-10-sulphonic acid, d-10-camphorsulfonic acid, 7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, l-camphor-10-sulfonic acid, +-10-camphorsulfonic acid
PubChem CID 18462
ChEBI CHEBI:55379
IUPAC Name (7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl)methanesulfonic acid
SMILES CC1(C2CCC1(C(=O)C2)CS(=O)(=O)O)C

Specifications

Absorbance (20% aq. soln.)
Melting Point >195°C
Color Beige
Loss on Drying 1.5% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 2.5 kg
Beilstein 11, 316
Fieser 14,71; 15,68; 16,58
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in glacial acetic acid,slightly soluble in ethyl acetate,practically insoluble in ether
Formula Weight 232.29
Clarity (5% in water) Clear solution
Percent Purity 98%
Physical Form Granular Powder
Chemical Name or Material DL-10-Camphorsulfonic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • DL-10-Camphorsulfonic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.