missing translation for 'onlineSavingsMsg'
Learn More

DL-10-Camphorsulfonic acid, sodium salt, 98%

Product Code. 10119981 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
500g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10119981 5 g Glass bottle 5g
10477560 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10119981 Supplier Thermo Scientific Acros Supplier No. 189120050
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 34850-66-3 | C10H15NaO4S | 254.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34850-66-3
Molecular Formula C10H15NaO4S
Molecular Weight (g/mol) 254.28
MDL Number MFCD00135623
InChI Key AWMAOFAHBPCBHJ-UHFFFAOYSA-M
Synonym sodium camphorsulfonate, sodium 7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonate, sodium +/--10-camphorsulfonate, d-camphor-10-sulfonic acid sodium salt, sodium camphorsulfonate anion, bicyclo 2.2.1 heptane-1-methanesulfonic acid, 7,7-dimethyl-2-oxo-, sodium salt, +--sodium camphorsulfonate, sodium 2-oxobornane-10-sulphonate, sodium di-10-camphorsulfonate
PubChem CID 23686666
IUPAC Name sodium;(7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl)methanesulfonate
SMILES CC1(C2CCC1(C(=O)C2)CS(=O)(=O)[O-])C.[Na+]

Specifications

Melting Point 286°C to 288°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble
Formula Weight 254.28
Percent Purity 98%
Physical Form Amorphous Powder
Chemical Name or Material DL-10-Camphorsulfonic acid, sodium salt, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.