missing translation for 'onlineSavingsMsg'
Learn More

DL-2,4-Diaminobutyric acid dihydrochloride, 99%

Product Code. 11428386
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11428386 25 g 25g
11408386 1 g 1g
11418386 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428386 Supplier Thermo Scientific Alfa Aesar Supplier No. A15408.14
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 65427-54-5 | C4H12Cl2N2O2 | 191.052 g/mol

2,4-Diaminobutyric Acid is also found to inhibit GABA transaminase, thus producing an elevation of GABA level, and to have an antitumor activityagainst mouse fibrosarcoma cells. It is also used as an internal standard for amino acid analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Diaminobutyric Acid is also found to inhibit GABA transaminase, thus producing an elevation of GABA level, and to have an antitumor activity against mouse fibrosarcoma cells. It is also used as an internal standard for amino acid analysis.

Solubility
Soluble in water 50 mg/mL.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65427-54-5
Molecular Formula C4H12Cl2N2O2
Molecular Weight (g/mol) 191.052
MDL Number MFCD00012910
InChI Key CKAAWCHIBBNLOJ-UHFFFAOYSA-N
Synonym dl-2,4-diaminobutyric acid dihydrochloride, 2,4-diaminobutanoic acid dihydrochloride, h-dl-dab.2hcl, l-2,4-diaminobutyric acid dihydrochloride, dl-2,4-diaminobutyricaciddihydrochloride, butanoic acid, 2,4-diamino-, dihydrochloride, l-2,4-diaminobutyric aicd dihydrochloride, 2,4-bis azanyl butanoic acid dihydrochloride, c4h10n2o2.2hcl, h-dl-dab-oh.2hcl
PubChem CID 2724329
IUPAC Name 2,4-diaminobutanoic acid;dihydrochloride
SMILES C(CN)C(C(=O)O)N.Cl.Cl

Specifications

Melting Point ∼198°C (decomposition)
Quantity 25 g
Beilstein 4708499
Sensitivity Hygroscopic
Solubility Information Soluble in water 50mg/mL.
Formula Weight 191.06
Percent Purity 99%
Chemical Name or Material DL-2,4-Diaminobutyric acid dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.