missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Amino-1,6-hexanedioic-2,5,5-d3 Acid, TRC

Product Code. 30316091
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316091 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30316091 Supplier Toronto Research Chemicals Supplier No. TRCA580501100MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-2-Amino-1,6-hexanedioic-2,5,5-d3 Acid
CAS 2483832-01-3
Molecular Formula C6 D3 H8 N O4
Quantity 100 mg
Synonym DL-2-Amino-1,6-hexanedioic-2,5,5-d3 Acid, 2-Aminohexanedioic-2,5,5-d3 acid, 2-Aminoadipic acid-2,5,5-d3, (±)-α-Aminoadipic acid-d3, DL-2-Aminoadipic-2,5,5-d3 acid, 2-Aminohexanedioic acid-2,5,5-d3, 2-Aminohexanedioic Acid-2,5,5-D3
InChI Formula InChI=1S/C6H11NO4/c7-4(6(10)11)2-1-3-5(8)9/h4H,1-3,7H2,(H,8,9)(H,10,11)/i3D2,4D
SMILES [2H]C([2H])(CCC([2H])(N)C(=O)O)C(=O)O
IUPAC Name 2-amino-2,5,5-trideuteriohexanedioic acid
Molecular Weight (g/mol) 164.1743
Formula Weight 164.0876
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.