missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Amino-3-methyl-1-butanol, 97%, Thermo Scientific Chemicals

Product Code. 10457190 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10457190 5 g 5g
10582201 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10457190 Supplier Thermo Scientific Acros Supplier No. 180420050
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16369-05-4 | Molecular Formula: C5H13NO | Molecular Weight: 103.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16369-05-4
MDL Number MFCD00004730
InChI Key NWYYWIJOWOLJNR-UHFFFAOYSA-N
Synonym 2-amino-3-methyl-1-butanol, dl-valinol, valinol, l-+-valinol, 2-amino-3-methyl-butan-1-ol, 1-butanol, 2-amino-3-methyl, 1-butanol, 2-amino-3-methyl-, .+/-., s-+-2-amino-3-methyl-1-butanol, 1-2-amino-3-methylbutan-1-ol
PubChem CID 79019
IUPAC Name 2-amino-3-methylbutan-1-ol
SMILES CC(C)C(CO)N

Spezifikation

Color Yellow
Density 0.9300g/mL
Boiling Point 75.0°C to 77.0°C (8.0 mmHg)
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C5H13NO
Linear Formula (CH3)2CHCH(NH2)CH2OH
Refractive Index 1.4533 to 1.4553
Quantity 5 g
Fieser 12,563; 13,341; 01,63; 02,32; 03,29; 04,41; 05,48; 09,59; 11,7
Specific Gravity 0.93
Molecular Weight (g/mol) 103.16
Formula Weight 103.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material DL-2-Amino-3-methyl-1-butanol, 95%
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.