missing translation for 'onlineSavingsMsg'
Learn More

Dl-2-amino-3-phenyl-1-propanol, 98%, Thermo Scientific™

Product Code. 10246842 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10246842 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10246842 Supplier Thermo Scientific Acros Supplier No. 374290010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 16088-07-6
Melting Point 64.0°C to 72.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C9H13NO
Linear Formula C6H5CH2CH(NH2)CH2OH
MDL Number MFCD00066689
Quantity 1 g
Beilstein 13, III, 1757
Synonym dl-phenylalaninol, 2-amino-3-phenyl-1-propanol, phenylalaninol, s-phenylalaninol, l-2-amino-3-phenyl-1-propanol, s-2-benzylethanolamine, benzenepropanol, beta-amino, s-beta-aminobenzenepropanol, chembl21676, 2-amino-3-phenyl-propan-1-ol
Solubility Information Solubility in water: soluble. Other solubilities: soluble in diethyl ether, benzene, petroleum ether
InChI Key STVVMTBJNDTZBF-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)CC(CO)N
IUPAC Name 2-amino-3-phenylpropan-1-ol
Molecular Weight (g/mol) 151.21
PubChem CID 76652
Formula Weight 151.21
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material DL-2-Amino-3-phenyl-1-propanol
Show More Show Less
compliance-icons
Product Identifier
  • DL-2-Amino-3-phenyl-1-propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.