missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Hydroxybutyric acid sodium salt, 97+%

Product Code. 11345229
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11345229 5 g 5g
11335229 1 g 1g
11355229 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11345229 Supplier Thermo Scientific Alfa Aesar Supplier No. A18636.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5094-24-6 | C4H7NaO3 | 126.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5094-24-6
Molecular Formula C4H7NaO3
Molecular Weight (g/mol) 126.09
MDL Number MFCD00004565
InChI Key MOSCXNXKSOHVSQ-UHFFFAOYNA-M
Synonym sodium 2-hydroxybutanoate, sodium 2-hydroxybutyrate, sodium dl-2-hydroxybutyrate, 2-hydroxybutyric acid sodium salt, dl-2-hydroxybutyric acid sodium salt, sodium hydroxybutyrate, acmc-1btqj, sodium, a-hydroxybutyrate, sodium 2-oxidanylbutanoate, sodiumdl-2-hydroxybutyrate
PubChem CID 23663641
IUPAC Name sodium;2-hydroxybutanoate
SMILES [Na+].CCC(O)C([O-])=O

Specifications

Melting Point 153°C to 157°C
Quantity 5 g
Beilstein 4825104
Sensitivity Hygroscopic
Formula Weight 126.09
Percent Purity ≥97%
Chemical Name or Material DL-2-Hydroxybutyric acid sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.