missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Pyrrolidone-5-carboxylic acid, 98%, Thermo Scientific™

Product Code. 10164570 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10164570 5 g Glass bottle 5g
10431851 100 g Plastic bottle 100g
10239213 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10164570 Supplier Thermo Scientific Acros Supplier No. 232930050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 149-87-1
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.12
MDL Number MFCD00064322
InChI Key ODHCTXKNWHHXJC-UHFFFAOYNA-N
Synonym dl-pyroglutamic acid, 2-pyrrolidone-5-carboxylic acid, dl-proline, 5-oxo, 5-oxo-dl-proline, pyroglutamate, h-dl-pyr-oh, pyrrolidonecarboxylic acid, 5-oxoprolinate, d-+-pyroglutamic acid, dl-pidolic acid
PubChem CID 499
ChEBI CHEBI:16010
IUPAC Name 5-oxopyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCC(=O)N1

Specifications

Melting Point 180.0°C to 185.0°C
Color White
Ignition Residue 0.3% max.
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: 5.67% (20°C). Other solubilities: soluble in alcohol and acetone
Formula Weight 129.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material DL-2-Pyrrolidone-5-carboxylic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • DL-2-Pyrrolidone-5-carboxylic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.