missing translation for 'onlineSavingsMsg'
Learn More

DL-3-Methyl-2-butanol, 98%

Product Code. 10427170 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10427170 25 mL Glass bottle 25mL
10497350 100 mL Glass bottle 100mL
10065763 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10427170 Supplier Thermo Scientific Acros Supplier No. 149930250
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 598-75-4 | C5H12O | 88.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-75-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004527
InChI Key MXLMTQWGSQIYOW-UHFFFAOYNA-N
Synonym 3-methyl-2-butanol, sec-isoamyl alcohol, 2-butanol, 3-methyl, 2-methyl-3-butanol, isopropylmethylcarbinol, methylisopropylcarbinol, 1,2-dimethylpropanol, 1,2-dimethyl-1-propanol, dl-3-methyl-2-butanol, 3-methyl-butan-2-ol
PubChem CID 11732
ChEBI CHEBI:77517
IUPAC Name 3-methylbutan-2-ol
SMILES CC(C)C(C)O

Specifications

Color Colorless
Density 0.8100g/mL
Boiling Point 112.0°C
Flash Point 39°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)2CHCH(OH)CH3
Refractive Index 1.4080 to 1.4100
Quantity 25 mL
Merck Index 15, 6105
Specific Gravity 0.81
Solubility Information Solubility in water: 28g/L (30°C). Other solubilities: miscible with alcohol and ether
Viscosity 3.8 mPa.s (25°C)
Formula Weight 88.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material DL-3-Methyl-2-butanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • DL-3-Methyl-2-butanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.