missing translation for 'onlineSavingsMsg'
Learn More

DL-6-Methyl-5-hepten-2-ol, 99%

Product Code. 10459680 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10459680 25 mL 25mL
10350994 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10459680 Supplier Thermo Scientific Acros Supplier No. 187690250
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4630-06-2 | C8H16O | 128.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6-2-4630
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.21
MDL Number MFCD00004561
InChI Key OHEFFKYYKJVVOX-UHFFFAOYNA-N
SMILES CC(O)CCC=C(C)C

Specifications

Color Colorless to Yellow
Density 0.8400g/mL
Boiling Point 78.0°C (14.0 mmHg)
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)2C=CHCH2CH2CH(OH)CH3
Refractive Index 1.4475 to 1.4495
Quantity 25 mL
Beilstein 01, IV, 2167
Specific Gravity 0.84
Synonym 6-methyl-5-hepten-2-ol, sulcatol, 5-hepten-2-ol, 6-methyl, r-sulcatol, 2-methyl-2-hepten-6-ol, +/--6-methyl-5-hepten-2-ol, 6-methyl-hept-5-en-2-ol, r---6-methyl-5-hepten-2-ol, 6-hydroxy-2-methyl-2-heptene, acmc-20apkw
IUPAC Name 6-methylhept-5-en-2-ol
PubChem CID 20745
ChEBI CHEBI:15833
Formula Weight 128.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material DL-6-Methyl-5-hepten-2-ol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.