missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Alaninol, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | DL-Alaninol |
| CAS | 6168-72-5 |
| Molecular Formula | C3H9NO |
| Quantity | 25 g |
| Synonym | (±)-2-Amino-1-propanol, DL-2-Amino-1-propanol, (RS)-2-Amino-1-propanol(±)-2-Amino-1-propanol, (±)-2-Aminopropanol, (±)-Alaninol, 1-Hydroxy-2-aminopropane, 1-Hydroxyprop-2-ylamine, 1-Hydroxypropan-2-amine1-Methyl-2-hydroxyethylamine, 2-Amino-1-hydroxypropane, 2-Amino-1-propanol2-Amino-2-methylethanol, 2-Aminopropanol, 2-Hydroxy-1-methylethylamine, AlaninolDL-2-Amino-1-propanol, NSC 1360, Propanolamine, [1-(Hydroxymethyl)ethyl]amine, dl-2-Amino-1-propanol, dl-2-Aminopropanol, dl-Alaninol, beta-Aminopropanol, beta-Propanolamine |
| InChI Formula | InChI=1S/C3H9NO/c1-3(4)2-5/h3,5H,2,4H2,1H3 |
| SMILES | CC(N)CO |
| IUPAC Name | 2-aminopropan-1-ol |
| Molecular Weight (g/mol) | 75.11 |
| Formula Weight | 75.0684 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction