missing translation for 'onlineSavingsMsg'
Learn More

DL-alpha-Lipoic Acid, ≥98% (Dry Basis)

Product Code. 15885168 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15885168 25 g Plastic bottle 25g
15895168 50 g Plastic bottle 50g
15805178 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15885168 Supplier Thermo Scientific Supplier No. J18505.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1077-28-7 | C8H14O2S2 | 206.318 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1077-28-7
Molecular Formula C8H14O2S2
Molecular Weight (g/mol) 206.318
MDL Number MFCD00005474
InChI Key AGBQKNBQESQNJD-UHFFFAOYSA-N
Synonym dl-thioctic acid, thioctic acid, alpha-lipoic acid, 5-1,2-dithiolan-3-yl pentanoic acid, dl-alpha-lipoic acid, 1,2-dithiolane-3-pentanoic acid, 6,8-thioctic acid, thioctacid, 6-thioctic acid, alpha lipoic acid
PubChem CID 864
ChEBI CHEBI:16494
IUPAC Name 5-(dithiolan-3-yl)pentanoic acid
SMILES C1CSSC1CCCCC(=O)O

Specifications

Melting Point 59°C to 63°C
Boiling Point 160°C to 165°C
Packaging Plastic bottle
Quantity 25 g
Beilstein 81853
Formula Weight 206.33
Percent Purity ≥98%
Physical Form Crystalline Powder
Assay Dry basis
Chemical Name or Material DL-alpha-Lipoic Acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.