missing translation for 'onlineSavingsMsg'
Learn More

dl-Alpha-Tocopherol Acetate, TRC

Product Code. 17987144
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17987144 10 g 10g
17979904 50 g 50g
17917714 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17987144 Supplier Toronto Research Chemicals Supplier No. TRCT52615510G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52225-20-4
Molecular Formula C31 H52 O3
Molecular Weight (g/mol) 472.74
Synonym 2H-1-Benzopyran-6-ol, 3,4-dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-, acetate, [2R*(4R*,8R*)]-(±)-, 2H-1-Benzopyran-6-ol, 3,4-dihydro-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-, acetate, (2R)-rel- (9CI), (±)-α-Tocopherol acetate, Alfacol, DL-α-Tocopherol acetate, DL-α-Tocopheryl acetate, Detulin, Diluvac Forte, E-Vimin, Ephynal, Eusovit, Evigen, Fertilvit, Gevex, OptoVit E, Rovimix E, Rovimix E 50, Rovimix E 50AS, Rovimix E 50Ads, Rovimix E 50SD, SynAC, Syntopherol acetate, Toco500, Tocopherol acetate, Vitagutt, all-rac-α-Tocopheryl acetate, dl-Vitamin E acetate, dl-α-Tocopherol acetate, dl-α-Tocopheryl acetate, α-Tocopherol acetate, α-Tocopheryl acetate
IUPAC Name [(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl] acetate
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2c(C)c(OC(=O)C)c(C)c(C)c2O1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C31H52O3/c1-21(2)13-10-14-22(3)15-11-16-23(4)17-12-19-31(9)20-18-28-26(7)29(33-27(8)32)24(5)25(6)30(28)34-31/h21-23H,10-20H2,1-9H3/t22-,23-,31-/m1/s1
Formula Weight 472.3916
Percent Purity >95%
Chemical Name or Material Vitamin E acetate (DL-alpha-Tocopherol acetate)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.