missing translation for 'onlineSavingsMsg'
Learn More

dl-Alpha-Tocopherol, TRC

Product Code. 30004959
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004959 50 g 50g
17963925 10 g 10g
17992215 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004959 Supplier Toronto Research Chemicals Supplier No. TRCT52614550G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10191-41-0
Molecular Formula C29 H50 O2
Molecular Weight (g/mol) 430.71
Synonym 6-Chromanol, 2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)- (8CI), 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-1-benzopyran-6-ol, (±)-α-Tocopherol, 2,5,7,8-Tetramethyl-2-(4',8',12'-trimethyltridecyl)chroman-6-ol, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, DL-α-Tocopherol, Elementol B, Elementol Basic, Elementol R, Ephanyl, Irganox E 210, Rac-α-Tocopherol, Ronotec 201, Ronotec 202, Ronotec DF 120, Uvinul 2000AO, all-rac-α-Tocopherol, dl-α-Tocopherol
IUPAC Name 2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-3,4-dihydrochromen-6-ol
SMILES CC(C)CCCC(C)CCCC(C)CCCC1(C)CCc2c(C)c(O)c(C)c(C)c2O1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C29H50O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h20-22,30H,9-19H2,1-8H3
Formula Weight 430.3811
Percent Purity >95%
Chemical Name or Material DL-alpha-Tocopherol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.