missing translation for 'onlineSavingsMsg'
Learn More

DL-α-Amino-n-butyric Acid, MP Biomedicals

Product Code. 11463110 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463110 500 g 500g
11443110 25 g 25g
11453110 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463110 Supplier MP Biomedicals Supplier No. 0210043090
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 56-12-2
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00008093
InChI Key BTCSSZJGUNDROE-UHFFFAOYSA-N
Synonym 4-aminobutyric acid, gaba, gamma-aminobutyric acid, piperidic acid, piperidinic acid, aminalon, gaballon, gammalon, mielogen, butanoic acid, 4-amino
PubChem CID 119
ChEBI CHEBI:16865
IUPAC Name 4-aminobutanoic acid
SMILES C(CC(=O)O)CN

Specifications

Quantity 500 g
Formula Weight 103.12
Chemical Name or Material DL-Alpha-Amino-n-Butyric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.