missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Arginine, 98%

Product Code. 11468137
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11468137 25 g 25g
11458137 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11468137 Supplier Thermo Scientific Chemicals Supplier No. A17520.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7200-25-1 | C6H14N4O2 | 174.204 g/mol

DL-Arginine is used in the synthesis of creatine and polyamines. DL-Arg is used in physicochemical analysis of amino acid complexation dynamics and crystal structure formations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Arginine is used in the synthesis of creatine and polyamines. DL-Arg is used in physicochemical analysis of amino acid complexation dynamics and crystal structure formations.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7200-25-1
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.204
MDL Number MFCD00063117
InChI Key ODKSFYDXXFIFQN-UHFFFAOYSA-N
Synonym dl-arginine, arginin, h-dl-arg-oh, 2-amino-5-carbamimidamidopentanoic acid, 2-amino-5-guanidinopentanoic acid, unii-fl26ntk3ep, dl-arginine, free base, fl26ntk3ep, wln: muyzm3yzvq &gh, arginine, dl
PubChem CID 232
ChEBI CHEBI:29016
IUPAC Name 2-amino-5-(diaminomethylideneamino)pentanoic acid
SMILES C(CC(C(=O)O)N)CN=C(N)N

Specifications

Melting Point ∼230°C (decomposition)
Beilstein 1725411
Solubility Information Insoluble in water.
Quantity 25 g
Formula Weight 174.2
Sensitivity Air Sensitive
Percent Purity 98%
Chemical Name or Material DL-Arginine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.