missing translation for 'onlineSavingsMsg'
Learn More

DL-Arginine hydrochloride, 98%, anhydrous, Thermo Scientific Chemicals

Product Code. 11995531 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11995531 1 g 1g
11905541 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11995531 Supplier Thermo Scientific Acros Supplier No. 446020010
Quantity Price
1 20.90 € / 1g
10 18.10 € / 1g

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 32042-43-6 | Molecular Formula: C6H14N4O2·ClH | Molecular Weight: 210.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32042-43-6
Molecular Formula C6H14N4O2·ClH
Molecular Weight (g/mol) 210.67
InChI Key KWTQSFXGGICVPE-UHFFFAOYSA-N
Synonym dl-arginine hydrochloride, 2-amino-5-guanidinopentanoic acid hydrochloride, argininhydrochlorid, dl-arginine hcl, h-dl-arg-oh.xhcl, dl-+/--arginine hydrochloride, 1-hydrochloride arginine, arginine monochloride, h-dl-arg-oh.hcl, wln: muyzm3yzvq &gh
PubChem CID 85880
IUPAC Name 2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN=C(N)N.Cl

Specifications

Melting Point 200.0°C to 203.0°C
Color White
Loss on Drying 3% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5%
Packaging Glass Bottle
Linear Formula H2NC(=NH)NH(CH2)3CH(NH2)CO2H·HCl
Quantity 1 g
Solubility Information Solubility in water: soluble.
Formula Weight 210.67
Percent Purity 98%
Physical Form Powder
Chemical Name or Material DL-Arginine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.