missing translation for 'onlineSavingsMsg'
Learn More

Dl-Arginine Hydrochloride, TRC

Product Code. 30374213
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30374213 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30374213 Supplier Toronto Research Chemicals Supplier No. TRCD5257831G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-Arginine hydrochloride
CAS 32042-43-6
Molecular Formula C6 H14 N4 O2 . Cl H
Quantity 1 g
Synonym Arginine, hydrochloride (9CI), DL-Arginine, hydrochloride, Arginine hydrochloride (1:?)
InChI Formula InChI=1S/C6H14N4O2.ClH/c7-4(5(11)12)2-1-3-10-6(8)9;/h4H,1-3,7H2,(H,11,12)(H4,8,9,10);1H
SMILES Cl.NC(CCCNC(=N)N)C(=O)O
IUPAC Name 2-amino-5-carbamimidamidopentanoic acid;hydrochloride
Molecular Weight (g/mol) 210.66
Formula Weight 210.0884
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.