missing translation for 'onlineSavingsMsg'
Learn More

DL-Aspartic acid, 99+%, MP Biomedicals™

Product Code. 11393979 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
Unit Size:
1kg
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393979 500 g 500g
11383979 250 g 250g
11303989 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11393979 Supplier MP Biomedicals Supplier No. 0210080390
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Racemic mixture of the proteinogenic amino acid L-aspartate and the non-proteinogenic amino acid D-aspartate, used in studies on semen and sperm quality.

  • CAS Number: 617-45-8
  • Molecular Formula: C4H7NO4
  • Molecular Weight: 133.1
  • Purity: ≥99.0%
  • Melting Point: >300 °C (Lit.)
  • Solubility: Soluble in NaOH 100 mg/ mL
  • pH: 3.0 (Lit.)
  • Optical Rotation: + 25.5 ° (Lit.)
  • Biochemical Physiological Actions: DL-Aspartic acid is a racemic mixture of the proteinogenic amino acid L-aspartate and the non-proteinogenic amino acid D-aspartate, used in studies on factors and conditions that enhance semen and sperm quality.
  • Usage Statement: Research or further manufacturing use only, not for direct human use.

Chemical Identifiers

CAS 617-45-8
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.10
MDL Number MFCD00063083
InChI Key CKLJMWTZIZZHCS-UHFFFAOYNA-N
Synonym dl-aspartic acid, 2-aminosuccinic acid, dl-aminosuccinic acid, dl-asparagic acid, aspartic acid, dl, dl-asp-oh, +--aspartic acid, r,s-aspartic acid, h-dl-asp-oh, acid d,l-aspart
PubChem CID 424
ChEBI CHEBI:22660
IUPAC Name 2-aminobutanedioic acid
SMILES NC(CC(O)=O)C(O)=O

Specifications

Assay Percent Range 99+%
Quantity 500 g
Formula Weight 133.1
Percent Purity ≥99.0%
Grade Research
Physical Form Crystalline
Chemical Name or Material DL-Aspartic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.