missing translation for 'onlineSavingsMsg'
Learn More

DL-Aspartic acid, 99+%

Product Code. 10385970 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic Bottle
Unit Size:
1kg
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10385970 5 g Glass Bottle 5g
10636672 250 g Plastic Bottle 250g
10721735 1 kg Plastic Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10385970 Supplier Thermo Scientific Acros Supplier No. 105030050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 617-45-8 | C4H7NO4 | 133.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-45-8
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.10
MDL Number MFCD00063083
InChI Key CKLJMWTZIZZHCS-UHFFFAOYNA-N
Synonym dl-aspartic acid, 2-aminosuccinic acid, dl-aminosuccinic acid, dl-asparagic acid, aspartic acid, dl, dl-asp-oh, +--aspartic acid, r,s-aspartic acid, h-dl-asp-oh, acid d,l-aspart
PubChem CID 424
ChEBI CHEBI:22660
IUPAC Name 2-aminobutanedioic acid
SMILES NC(CC(O)=O)C(O)=O

Specifications

Ammonium 500ppm max.
Color Yellow
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 99+%
Loss on Drying 0.3% max. (105°C, 3 hrs)
Packaging Glass Bottle
Linear Formula HO2CCH2CH(NH2)CO2H
Beilstein 04, 483
Specific Rotation −0.3° to +0.3° (20°C, 589nm) (c=10, 2 N HCl)
Sulfate 500ppm max.
Merck Index 10, 853
Transmittance 95% min. (c=10, 2 N HCl, 430nm) 1cm cell
Sulfated Ash 0.1% max.
Quantity 5 g
Formula Weight 133.1
Percent Purity 99.0 to 101.0% (Acid)
Physical Form Crystalline Low Melting Mass
Chemical Name or Material DL-Aspartic acid, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.