missing translation for 'onlineSavingsMsg'
Learn More

DL-beta-Prolinol, 97+%, Thermo Scientific Chemicals

Product Code. 15453748
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15453748

Brand: Thermo Scientific Alfa Aesar H57225.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5082-74-6
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD07773080
InChI Key QOTUIIJRVXKSJU-UHFFFAOYNA-N
Synonym 3-hydroxymethylpyrrolidine, 3-pyrrolidinemethanol, pyrrolidin-3-yl-methanol, pyrrolidine-3-yl-methanol, pyrrolidin-3-yl methanol, 3-pyrrolidinylmethanol, dl-beta-prolinol, rs-3-pyrrolidinemethanol, s-3-hydroxymethyl pyrrolidine, 3-pyrrolidinemethanol, 3s
PubChem CID 13400657
IUPAC Name (pyrrolidin-3-yl)methanol
SMILES OCC1CCNC1

Specifications

Density 0.979
Boiling Point 165°C to 171°C
Quantity 250 mg
Sensitivity Hygroscopic
Beilstein 1420904
Solubility Information Slightly soluble in water
Formula Weight 101.15
Percent Purity ≥97%
Chemical Name or Material DL-beta-Prolinol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.