missing translation for 'onlineSavingsMsg'
Learn More

DL-beta-Prolinol, 97+%, Thermo Scientific Chemicals

Product Code. p-7038300
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15453748 250 mg 250mg
15443748 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15453748 Supplier Thermo Scientific Alfa Aesar Supplier No. H57225.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5082-74-6
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD07773080
InChI Key QOTUIIJRVXKSJU-UHFFFAOYNA-N
Synonym 3-hydroxymethylpyrrolidine, 3-pyrrolidinemethanol, pyrrolidin-3-yl-methanol, pyrrolidine-3-yl-methanol, pyrrolidin-3-yl methanol, 3-pyrrolidinylmethanol, dl-beta-prolinol, rs-3-pyrrolidinemethanol, s-3-hydroxymethyl pyrrolidine, 3-pyrrolidinemethanol, 3s
PubChem CID 13400657
IUPAC Name (pyrrolidin-3-yl)methanol
SMILES OCC1CCNC1

Specifica

Density 0.979
Boiling Point 165°C to 171°C
Quantity 250 mg
Sensitivity Hygroscopic
Beilstein 1420904
Solubility Information Slightly soluble in water
Formula Weight 101.15
Percent Purity ≥97%
Chemical Name or Material DL-beta-Prolinol

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.