missing translation for 'onlineSavingsMsg'
Learn More

DL-alpha-Bromophenylacetic acid, 97%

Product Code. 11904981 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11904981 5 g Glass bottle 5g
11914981 25 g Glass bottle 25g
10102950 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11904981 Supplier Thermo Scientific Acros Supplier No. 107270050
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 4870-65-9 | C8H7BrO2 | 215.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4870-65-9
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.04
InChI Key WAKFRZBXTKUFIW-UHFFFAOYSA-N
Synonym alpha-bromophenylacetic acid, bromo phenyl acetic acid, .alpha.-bromophenylacetic acid, bromophenylacetic acid, bromo-phenyl-acetic acid, bromo-phenylacetic acid, dl-.alpha.-bromophenylacetic acid, benzeneacetic acid, .alpha.-bromo, phenylbromoacetic acid, bromo phenyl aceticacid
PubChem CID 97919
IUPAC Name 2-bromo-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)Br

Specifications

Melting Point 82.0°C to 83.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH(Br)CO2H
Quantity 5 g
Beilstein 09,451
Solubility Information Solubility in water: insoluble
Formula Weight 215.04
Percent Purity 97%
Chemical Name or Material DL-α-Bromophenylacetic acid
compliance-icons
Product Identifier
  • DL-alpha-Bromophenylacetic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.