missing translation for 'onlineSavingsMsg'
Learn More

DL-Camphor, 96%

Product Code. 10127653 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
250g
5kg
This item is not returnable. View return policy

Product Code. 10127653

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 76-22-2 | C10H16O | 152.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-22-2
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00074738,MFCD00064149
InChI Key DSSYKIVIOFKYAU-UHFFFAOYNA-N
Synonym camphor, dl-camphor, 2-camphanone, 2-bornanone, l---camphor, +/--camphor, root bark oil, 1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, bornan-2-one, alphanon
PubChem CID 2537
ChEBI CHEBI:36773
IUPAC Name 4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
SMILES CC1(C)C2CCC1(C)C(=O)C2

Specifications

Melting Point 175°C to 177°C
Density 64g/mL
Boiling Point 204°C
Flash Point 64°C
Assay Percent Range 95% min. (GC)
Packaging Plastic bottle
Quantity 250 g
Beilstein 07,IV,213
Merck Index 15,1734
Solubility Information Solubility in water: 1.5g/L (20°C). Other solubilities: at 25°C 1 g dissolves in 1 mL alcohol,1 mL ether,,0.5 mL chloroform,0.4 mL benzene,0.4 mL acetone,,1.5 mL turpentine oil,0.5 mL glacial acetic acid.,soluble in aniline,nitrobenzene,c
Formula Weight 152.24
Percent Purity 96%
Chemical Name or Material DL-Camphor
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • DL-Camphor
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 2
Hazard Statement
  • H228-Flammable solid.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H371-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
Supplemental information
  • MIXTURE LIST-Contains : Camphor

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.