missing translation for 'onlineSavingsMsg'
Learn More

DL-Cysteine hydrochloride, 95%, Thermo Scientific™

Product Code. 15274238 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15274238 5 g 5g
15284238 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15274238 Supplier Thermo Scientific Acros Supplier No. 448260050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 10318-18-0
Molecular Formula C3H8ClNO2S
Molecular Weight (g/mol) 157.61
MDL Number MFCD00064552,MFCD00012632
InChI Key IFQSXNOEEPCSLW-UHFFFAOYNA-N
Synonym dl-cysteine hydrochloride, 2-amino-3-mercaptopropanoic acid hydrochloride, cysteine hydrochloride, +--cysteine hydrochloride, dl-cysteine hcl, h-dl-cys-oh.hcl, cysteine, hydrochloride, dl, dl-cysteine hydrochloride hydrate, dl-cysteine hydrochloride anhydrous, 2-amino-3-sulfanylpropanoic acid hydrochloride
PubChem CID 25150
IUPAC Name 2-amino-3-sulfanylpropanoic acid;hydrochloride
SMILES [H+].[Cl-].NC(CS)C(O)=O

Specifications

Melting Point 156.0°C to 161.0°C
Infrared Spectrum Authentic
Loss on Drying 1% max. Vacuum, room temp, 20 hours+
Linear Formula HSCH2CH(NH2)CO2H·HCl
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and acetone
Quantity 5 g
Formula Weight 157.61
Percent Purity ≥94.0% (HPLC)
Chemical Name or Material DL-Cysteine hydrochloride
compliance-icons
Product Identifier
  • DL-Cysteine hydrochloride, anhydrous, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.