missing translation for 'onlineSavingsMsg'
Learn More

DL-Cysteine hydrochloride, 90+%

Product Code. 11491130
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491130 100 g 100g
11471130 5 g 5g
11481130 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491130 Supplier Thermo Scientific Alfa Aesar Supplier No. H25779.22
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10318-18-0 | C3H8ClNO2S | 157.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10318-18-0
Molecular Formula C3H8ClNO2S
Molecular Weight (g/mol) 157.61
MDL Number MFCD00064552,MFCD00012632
InChI Key IFQSXNOEEPCSLW-UHFFFAOYNA-N
Synonym dl-cysteine hydrochloride, 2-amino-3-mercaptopropanoic acid hydrochloride, cysteine hydrochloride, +--cysteine hydrochloride, dl-cysteine hcl, h-dl-cys-oh.hcl, cysteine, hydrochloride, dl, dl-cysteine hydrochloride hydrate, dl-cysteine hydrochloride anhydrous, 2-amino-3-sulfanylpropanoic acid hydrochloride
PubChem CID 25150
IUPAC Name 2-amino-3-sulfanylpropanoic acid;hydrochloride
SMILES [H+].[Cl-].NC(CS)C(O)=O

Specifications

Melting Point 140°C to 142°C
Beilstein 4150473
Quantity 100 g
Formula Weight 157.62
Sensitivity Air sensitive; Hygroscopic
Percent Purity ≥98%
Chemical Name or Material DL-Cysteine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.