missing translation for 'onlineSavingsMsg'
Learn More

DL-Cysteine, TRC

Product Code. 30365182
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30365182 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30365182 Supplier Toronto Research Chemicals Supplier No. TRCC99501510G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-Cysteine
CAS 3374-22-9
Molecular Formula C3 H7 N O2 S
Quantity 10 g
Synonym Cysteine, DL- (8CI), DL-Cysteine, (±)-Cysteine, NSC 63864
InChI Formula InChI=1S/C3H7NO2S/c4-2(1-7)3(5)6/h2,7H,1,4H2,(H,5,6)
SMILES NC(CS)C(=O)O
IUPAC Name 2-amino-3-sulfanylpropanoic acid
Molecular Weight (g/mol) 121.16
Formula Weight 121.0197
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.