missing translation for 'onlineSavingsMsg'
Learn More

DL-Dithiothreitol, MP Biomedicals™

Product Code. 11445832 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445832 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11445832 Supplier MP Biomedicals Supplier No. 04856126
  • ONLINE EXCLUSIVE PRICE
    250.00 €
    236.25 € / 5g
    Save 13.75 €
    5% Off
 Restricted Item
Estimated Shipment: 05-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

DL-Dithiothreitol, Electrophoresis grade, ≥99.5%, White Powder

  • Dithiothreitol is useful for stabilizing sulfhydryl containing enzymes.
  • Effective in sample buffers for reducing protein disulfide bonds prior to SDS-PAGE.
  • DTT can also be used for reducing the disulfide bridge of the cross-linker N,N'-bis(acryloyl)cystamine to break apart the matrix of a polyacrylamide gel.
  • DTT is less pungent and is less toxic than 2-mercaptoethanol.
  • DL-Dithiothreitol is also known as Clelands reagent; Protective agent for sulfhydryl groups (-SH). Quantitatively reduces disulfides (-S-S- to -SH).
  • In this reaction the DTT is oxidized to the cyclic disulfide which ensures the reduction of other disulfides in solution.

Chemical Identifiers

CAS 12-3-3483
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
MDL Number MFCD00004877
InChI Key VHJLVAABSRFDPM-IMJSIDKUSA-N
Synonym 1,4-Dithiothreitol, DTT, 1,4-Dithio-DL-threitol, Cleland Reagent

Specifications

Melting Point 40°C to 50°C
Color White
pH 4.0 to 6.0 (0.1 M in H2O, 25°C)
Flash Point 113°C / 235.4°F (lit.)
Quantity 5 g
UN Number UN3335
Beilstein 1719757
Solubility Information Soluble in water (50mg/mL - clear and colorless solution); soluble in ethanol, acetone, ethylate, chloroform and ether.
Percent Purity 99.5%
Grade ≥99.5%
Physical Form Powder
Show More Show Less
compliance-icons
Product Identifier
  • DL-Dithiothreitol, electrophoresis grade
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: DL-Dithiothreitol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.