missing translation for 'onlineSavingsMsg'
Learn More

DL-Ethyl 2-bromopropionate, 99%

Product Code. 10784031 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10784031 100 g Glass bottle 100g
10234670 250 g Glass bottle 250g
10399313 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10784031 Supplier Thermo Scientific Acros Supplier No. 118141000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 535-11-5 | C5H9BrO2 | 181.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 535-11-5
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00000144
InChI Key ARFLASKVLJTEJD-UHFFFAOYNA-N
Synonym ethyl 2-bromopropionate, ethyl alpha-bromopropionate, ethyl-2-bromopropionate, 2-bromopropanoic acid ethyl ester, propanoic acid, 2-bromo-, ethyl ester, alpha-bromopropionic acid ethyl ester, ethyl a-bromopropionate, dl-ethyl 2-bromopropionate, 2-bromopropionic acid ethyl ester, ethyl alpha-bromopropanoate
PubChem CID 79040
IUPAC Name ethyl 2-bromopropanoate
SMILES CCOC(=O)C(C)Br

Specifications

Color Colorless to Yellow
Density 1.3940g/mL
Boiling Point 156°C to 160°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(Br)CO2C2H5
Refractive Index 1.4450 to 1.447
Quantity 100 g
Beilstein 02,255
Merck Index 15,3827
Specific Gravity 1.394
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol and ether
Formula Weight 181.03
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material DL-Ethyl 2-bromopropionate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.