missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Glutamic acid monohydrate, 98+%

Product Code. 10086233 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10086233 25 g Plastic bottle 25g
10316800 100 g Plastic bottle 100g
10489410 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10086233 Supplier Thermo Scientific Chemicals Supplier No. 119930250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19285-83-7 | C5H9NO4·H2O | 165.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19285-83-7
Molecular Formula C5H9NO4·H2O
Molecular Weight (g/mol) 165.15
MDL Number MFCD00150703
InChI Key OZDAOHVKBFBBMZ-UHFFFAOYSA-N
Synonym dl-glutamic acid monohydrate, 2-aminopentanedioic acid hydrate, dl-glutamic acid hydrate, h-dl-glu-oh.h2o, glutaminsaeure hydrate, +/--2-aminoglutaric acid, dl-2-aminopentanedioic acid, 2-azanylpentanedioic acid hydrate, 2-aminopentanedioic acid, oxamethane, hydrate, a-aminoglutarate
PubChem CID 16219385
IUPAC Name 2-aminopentanedioic acid;hydrate
SMILES C(CC(=O)O)C(C(=O)O)N.O

Specifications

Melting Point 194.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Linear Formula HO2CCH2CH2CH(NH2)CO2H·H2O
Quantity 25 g
Solubility Information Solubility in water: 20.54g/L (25°C). Other solubilities: sparingly soluble in alcohol and ether
Formula Weight 165.15
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material DL-Glutamic acid monohydrate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.