missing translation for 'onlineSavingsMsg'
Learn More

DL-Glutamic Acid, TRC

Product Code. 30404189
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404189 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30404189 Supplier Toronto Research Chemicals Supplier No. TRCG59695550G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-Glutamic acid
CAS 617-65-2
Molecular Formula C5 H9 N O4
Quantity 50 g
Synonym Glutamic acid (9CI, ACI), DL-Glutamic acid (ZCI), Glutamic acid, DL- (8CI), (±)-Glutamic acid, 2-Aminopentanedioic acid, 4-Amino-1,5-pentanedioic acid, DL-Glu, Glutaminic acid, NSC 206301, NSC 9967
InChI Formula InChI=1S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)
SMILES NC(CCC(=O)O)C(=O)O
IUPAC Name 2-aminopentanedioic acid
Molecular Weight (g/mol) 147.13
Formula Weight 147.0532
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.