missing translation for 'onlineSavingsMsg'
Learn More

DL-Glutamine, 98%

Product Code. 15499358
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499358 5 g 5g
15489358 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499358 Supplier Thermo Scientific Alfa Aesar Supplier No. H62915.06
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6899-04-3 | C5H10N2O3 | 146.146 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6899-04-3
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.146
MDL Number MFCD00065103
InChI Key ZDXPYRJPNDTMRX-UHFFFAOYSA-N
Synonym dl-glutamine, glutamin, 2-amino-4-carbamoylbutanoic acid, dl glutamine, glutamine dl-form, glutamine, dl, l-2-aminoglutaramic acid, 2,5-bis azanyl-5-oxidanylidene-pentanoic acid, gamma-glutamine, .gamma.-glutamine
PubChem CID 738
ChEBI CHEBI:28300
IUPAC Name 2,5-diamino-5-oxopentanoic acid
SMILES C(CC(=O)N)C(C(=O)O)N

Specifications

Melting Point 177°C
Merck Index 14,4471
Quantity 5 g
Formula Weight 146.14
Percent Purity 98%
Chemical Name or Material DL-Glutamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.