missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Histidine monohydrochloride monohydrate, 99%

Product Code. 11423227
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423227 100 g 100g
11403227 5 g 5g
11413227 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423227 Supplier Thermo Scientific Chemicals Supplier No. A16165.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123333-71-1 | C6H12ClN3O3 | 209.63 g/mol

DL-Histidine monohydrochloride monohydrate is the mixed isomer of D- and L-histidine. It may be used to evaluate the efficiency of amino acid chiral ligand exchange media and systems and in running buffers for capillary electrophoresis. It is also an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Histidine monohydrochloride monohydrate is the mixed isomer of D- and L-histidine. It may be used to evaluate the efficiency of amino acid chiral ligand exchange media and systems and in running buffers for capillary electrophoresis. It is also an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123333-71-1
Molecular Formula C6H12ClN3O3
Molecular Weight (g/mol) 209.63
MDL Number MFCD00151028
InChI Key CMXXUDSWGMGYLZ-UHFFFAOYSA-N
Synonym dl-histidine; hypochlorous acid
PubChem CID 44828786
IUPAC Name 2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydron;chloride;hydrate
SMILES [H+].C1=C(NC=N1)CC(C(=O)O)N.O.[Cl-]

Specifications

Melting Point ∼250°C (decomposition)
Solubility Information Insoluble in water.
Quantity 100 g
Formula Weight 209.63 (191.62 Anhydrous)
Percent Purity 99%
Chemical Name or Material DL-Histidine monohydrochloride monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.