missing translation for 'onlineSavingsMsg'
Learn More

DL-Homocysteine, 90%, TRC

Product Code. 17942565
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17942565 250 mg 250mg
17911005 50 mg 50mg
17974895 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17942565 Supplier Toronto Research Chemicals Supplier No. TRCH591295250MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 454-29-5
Molecular Formula C4 H9 N O2 S
Molecular Weight (g/mol) 135.19
Synonym 2-Amino-4-mercaptobutyric acid
IUPAC Name 2-amino-4-sulfanyl-butanoic acid
SMILES NC(CCS)C(=O)O

Specifications

InChI Formula InChI=1S/C4H9NO2S/c5-3(1-2-8)4(6)7/h3,8H,1-2,5H2,(H,6,7)
Quantity 250 mg
Formula Weight 135.0354
Shipping Condition Room Temperature
Chemical Name or Material DL-Homocysteine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.