missing translation for 'onlineSavingsMsg'
Learn More

DL-Homocysteine thiolactone hydrochloride, 99%

Product Code. 11357216
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357216 25 g 25g
11367216 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357216 Supplier Thermo Scientific Alfa Aesar Supplier No. L09077.14
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 6038-19-3 | C4H8ClNOS | 153.624 g/mol

DL-Homocysteine thiolactone hydrochloride is used as an intermediate of erdosteine and citiolone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Homocysteine thiolactone hydrochloride is used as an intermediate of erdosteine and citiolone.

Solubility
Soluble in water (740g/L). Soluble in dimethyl sulfoxide and methanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6038-19-3
Molecular Formula C4H8ClNOS
Molecular Weight (g/mol) 153.624
MDL Number MFCD00012724
InChI Key ZSEGSUBKDDEALH-UHFFFAOYSA-N
Synonym dl-homocysteine thiolactone hydrochloride, 3-aminodihydrothiophen-2 3h-one hydrochloride, 3-aminothiolan-2-one hydrochloride, hctl hydrochloride, dl-homocysteinethiolactone hydrochloride, d,l-homocysteine thiolactone hydrochloride, d,l-homocysteinthiolakton chlorid german, +--dihydro-3-amino-2 3h-thiophenone hydrochloride, 2 3h-thiophenone, dihydro-3-amino-, hydrochloride, +-, l-2-amino-4-mercaptobutyric acid 1,4-thiolactone hydrochloride
PubChem CID 110753
IUPAC Name 3-aminothiolan-2-one;hydrochloride
SMILES C1CSC(=O)C1N.Cl

Specifications

Melting Point ∽204°C (decomposition)
Quantity 25 g
Beilstein 3562103
Merck Index 14,4734
Solubility Information Soluble in water (740g/L). Soluble in dimethyl sulfoxide and methanol.
Formula Weight 153.63
Percent Purity 99%
Chemical Name or Material DL-Homocysteine thiolactone hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.