missing translation for 'onlineSavingsMsg'
Learn More

dl Homocysteine Thiolactone Hydrochloride, TRC

Product Code. 30398485
Change view
Click to view available options
Quantity:
10 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30398485 250 g
30396393 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30398485 Supplier Toronto Research Chemicals Supplier No. TRCH591300250G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6038-19-3
Molecular Formula C4 H7 N O S . Cl H
Molecular Weight (g/mol) 153.63
Synonym 2(3H)-Thiophenone, 3-aminodihydro-, hydrochloride (1:1) (ACI), 2(3H)-Thiophenone, 3-aminodihydro-, hydrochloride (9CI), 2(3H)-Thiophenone, 3-aminodihydro-, hydrochloride, (±)- (8CI), 2-Oxothiolan-3-aminium chloride, 3-Aminodihydrothiophen-2(3H)-one hydrochloride, DL-Homocysteine thiolactone hydrochloride, ADT, HCTL hydrochloride, Homocysteine thiolactone hydrochloride, NSC 22879
IUPAC Name 3-aminothiolan-2-one;hydrochloride
SMILES Cl.NC1CCSC1=O

Specifications

InChI Formula InChI=1S/C4H7NOS.ClH/c5-3-1-2-7-4(3)6;/h3H,1-2,5H2;1H
Quantity 250 g
Formula Weight 153.0015
Shipping Condition Room Temperature
Chemical Name or Material DL-Homocysteine thiolactone hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.