missing translation for 'onlineSavingsMsg'
Learn More

DL-Homocystine, MedChemExpress

Product Code. 16402678
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16402678 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16402678 Supplier Medchem Express Supplier No. HYW009390
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress DL-Homocystine is the double-bonded form of homocysteine and homocysteine is recognized as an important substance in the pathogenesis and pathophysiology of schizophrenia.

DL-Homocystine is the double-bonded form of homocysteine and homocysteine is recognized as an important substance in the pathogenesis and pathophysiology of schizophrenia.

Specifications

Chemical Name or Material DL-Homocystine
CAS 870-93-9
Color White
Purity Grade Notes Research
Molecular Formula C8H16N2O4S2
Quantity 5 g
Solubility Information H2O : 10 mg/mL (37.26 mM; ultrasonic and adjust pH to 11 with NaOH)
SMILES N[C@H](C(O)=O)CCSSCC[C@H](N)C(O)=O.N[C@@H](C(O)=O)CCSSCC[C@@H](N)C(O)=O
Molecular Weight (g/mol) 268.35
Formula Weight 268.35
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.