missing translation for 'onlineSavingsMsg'
Learn More

DL-Lactic Acid (90%), TRC

Product Code. 30132466
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
100g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132466 500 g 500g
30138078 100 g 100g
30005784 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132466 Supplier Toronto Research Chemicals Supplier No. TRCL113490500G
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-21-5
Molecular Formula C3 H6 O3
Molecular Weight (g/mol) 90.08
Synonym Lactic acid, Propanoic acid, 2-hydroxy-, Lactic acid (7CI,8CI), 2-Hydroxypropanoic acid, (RS)-2-Hydroxypropanoic acid, (±)-Lactic acid, 2-Hydroxy-2-methylacetic acid, 2-Hydroxypropionic acid, Biolac, Chem-Cast, DL-Lactic acid, E 270, HiPure 88, Kiwilustre, Lurex, Masashino Lactic acid 90F, Milk acid, Musashino lactate 50F, NSC 367919, Purac 55, Purac 60, Purac FCC 80, Purac FCC 88, Purac HS 100, Purac HS 50, Purac HS 88, Tonsillosan, dl-Lactic acid, α-Hydroxypropanoic acid, α-Hydroxypropionic acid
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(=O)O

Specifications

InChI Formula InChI=1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)
Formula Weight 90.0317
Quantity 500 g
Chemical Name or Material DL-Lactic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.