missing translation for 'onlineSavingsMsg'
Learn More

DL-Lactic Acid Lithium Salt, MP Biomedicals™

Product Code. 11384009 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11384009 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11384009 Supplier MP Biomedicals Supplier No. 0210082405
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 867-55-0
Color White
Quantity 5 g
Physical Form Powder
Molecular Formula C3H5LiO3
MDL Number MFCD00065399
Synonym lithium lactate, lithium 2-hydroxypropanoate, lactic acid, lithium salt, dl-lactic acid lithium salt, propanoic acid, 2-hydroxy-, monolithium salt, lithium 1+ ion lactate, l-+-lactic acid lithium salt, lithotab lactate, acmc-20akmj, lithium s-lactate
InChI Key GKQWYZBANWAFMQ-UHFFFAOYSA-M
SMILES [Li+].CC(C(=O)[O-])O
IUPAC Name lithium;2-hydroxypropanoate
Molecular Weight (g/mol) 96.01
PubChem CID 23673452
Formula Weight 96.01
Chemical Name or Material DL-Lactic Acid Lithium Salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.