missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Leucine, 99%

Product Code. 11483254
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483254 50 g 50g
11493254 250 g 250g
11473254 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11483254 Supplier Thermo Scientific Chemicals Supplier No. A10590.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 328-39-2 | C6H13NO2 | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 328-39-2
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00063087
InChI Key ROHFNLRQFUQHCH-UHFFFAOYNA-N
Synonym dl-leucine, h-dl-leu-oh, leucine, dl, rs-leucine, +--leucine, dl-2-amino-4-methylpentanoic acid, 2-amino-4-methyl-pentanoic acid, l-1-13c leucine, l-1-14c leucine, leucine,d
PubChem CID 857
ChEBI CHEBI:25017
IUPAC Name 2-amino-4-methylpentanoic acid
SMILES CC(C)CC(N)C(O)=O

Specifications

Melting Point ∼294°C (sublimation)
Beilstein 636005
Merck Index 14,5451
Density 1.293
Quantity 50 g
Formula Weight 131.18
Percent Purity 99%
Chemical Name or Material DL-Leucine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.