missing translation for 'onlineSavingsMsg'
Learn More

DL-Lysine monohydrate, 97%

Product Code. 10691073 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10691073 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10691073 Supplier Thermo Scientific Acros Supplier No. 365650250
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 885701-25-7 | C6H16N2O3 | 164.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 885701-25-7
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Loss on Drying 7% to 12% (105°C, 2 hrs)
Molecular Formula C6H16N2O3
Packaging Glass Bottle
Linear Formula HN(CH)4CH(NH)COH
MDL Number MFCD00151036
Synonym dl-lysine monohydrate, 2,6-diaminohexanoic acid hydrate, lysine hydrate, l-2,6-diaminohexanoic acid, dl-lysine hydrate, 2,6-diaminohexanoic acid,hydrate
InChI Key HZRUTVAFDWTKGD-UHFFFAOYNA-N
SMILES O.NCCCCC(N)C(O)=O
IUPAC Name 2,6-diaminohexanoic acid;hydrate
Molecular Weight (g/mol) 164.21
Quantity 25 g
PubChem CID 2734605
Formula Weight 164.21
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material DL-Lysine monohydrate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.