missing translation for 'onlineSavingsMsg'
Learn More

Dl-lysine Monohydrate, TRC

Product Code. 30482175
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482175 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30482175 Supplier Toronto Research Chemicals Supplier No. TRCL3303051G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dl-lysine monohydrate
CAS 885701-25-7
Molecular Formula C6 H14 N2 O2 . H2 O
Quantity 1 g
Synonym Lysine, hydrate (1:1) (ACI), Lysine hydrate (1:1) (ACI), Lysine, monohydrate (9CI), 2,6-Diaminohexanoic acid hydrate, DL-Lysine hydrate, DL-Lysine monohydrate, 2,6-Diaminohexanoic acid monohydrate
InChI Formula InChI=1S/C6H14N2O2.H2O/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H2
SMILES O.NCCCCC(N)C(=O)O
IUPAC Name 2,6-diaminohexanoic acid;hydrate
Molecular Weight (g/mol) 164.2
Formula Weight 164.1161
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.