missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Lysine monohydrochloride, 99%

Product Code. 11428923
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428923 25 g 25g
11438923 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11428923 Supplier Thermo Scientific Chemicals Supplier No. A11066.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 70-53-1 | C6H15ClN2O2 | 182.65 g/mol

DL-Lysine monohydrochloride acts as an intermediate in the preparation of 3-amino-azepan-2-one. It also serves as an essential amino acid. It is used as animal feed. In addition to this, it is used in the construction of polylysine polymers, which finds application in lysine-based polypeptide gels and films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Lysine monohydrochloride acts as an intermediate in the preparation of 3-amino-azepan-2-one. It also serves as an essential amino acid. It is used as animal feed. In addition to this, it is used in the construction of polylysine polymers, which finds application in lysine-based polypeptide gels and films.

Solubility
Soluble in water. Insoluble in ethanol, ethyl ether, acetone, benzene and common neutral solvents.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-53-1
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00064563
InChI Key BVHLGVCQOALMSV-UHFFFAOYSA-N
Synonym dl-lysine monohydrochloride, 2,6-diaminohexanoic acid hydrochloride, dl-lysine hydrochloride, lysine monohydrochloride, dl-lysine mono hydrochloride, lysine, hydrochloride 1:1, lysine, monohydrochloride, dl-2,6-diaminohexanoic acid monohydrochloride, lysine, dl, dl-lysinemonohydrochloride
PubChem CID 12047
IUPAC Name 2,6-diaminohexanoic acid;hydrochloride
SMILES Cl.NCCCCC(N)C(O)=O

Specifications

Melting Point 267°C (decomposition)
Beilstein 4711993
Solubility Information Soluble in water. Insoluble in ethanol,ethyl ether,acetone,benzene and common neutral solvents.
Quantity 25 g
Formula Weight 182.65
Percent Purity 99%
Chemical Name or Material DL-Lysine monohydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.