missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-m-Tyrosine, 98.5%

Product Code. 10153620 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10153620 1 g Glass Bottle 1g
10637294 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10153620 Supplier Thermo Scientific Chemicals Supplier No. 140630010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 775-06-4 | C9H11NO3 | 181.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 775-06-4
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00002597
InChI Key JZKXXXDKRQWDET-UHFFFAOYNA-N
Synonym dl-m-tyrosine, 2-amino-3-3-hydroxyphenyl propanoic acid, 3-3-hydroxyphenyl-dl-alanine, m-tyrosine, dl, d,l-metatyrosine, 3-hydroxy-dl-phenylalanine, dl-meta-tyrosine, dl-phenylalanine, 3-hydroxy, 3-m-hydroxyphenyl-dl-alanine, dl-3-tyrosine
PubChem CID 13052
IUPAC Name 2-amino-3-(3-hydroxyphenyl)propanoic acid
SMILES NC(CC1=CC=CC(O)=C1)C(O)=O

Specifications

Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.50%
Packaging Glass Bottle
Linear Formula HOC6H4CH2CH(NH2)CO2H
Beilstein 14, 605
Merck Index 14, 9840
Water 0.5% max. (K.F.)
Quantity 1 g
Formula Weight 181.19
Percent Purity ≥98.0%
Physical Form Crystalline Powder
Chemical Name or Material DL-m-Tyrosine
Show More Show Less
compliance-icons
Product Identifier
  • DL-m-Tyrosine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.