missing translation for 'onlineSavingsMsg'
Learn More

DL-Malic Acid, BAKER ANALYZED™ Biochemical Reagent, J.T. Baker™

Product Code. 15548814 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15548814 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15548814 Supplier Avantor J.T.Baker Supplier No. P49407
 Restricted Item
Estimated Shipment: 10-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality biochemicals for research use

Specifications

CAS 617-48-1
Molecular Formula C4H6O5
Synonym malic acid, dl-malic acid, 2-hydroxysuccinic acid, malate, hydroxysuccinic acid, butanedioic acid, hydroxy, kyselina jablecna, pomalus acid, hydroxybutanedioic acid, deoxytetraric acid
InChI Key BJEPYKJPYRNKOW-UHFFFAOYSA-N
SMILES C(C(C(=O)O)O)C(=O)O
IUPAC Name 2-hydroxybutanedioic acid
Molecular Weight (g/mol) 134.087
PubChem CID 525
ChEBI CHEBI:6650
Formula Weight 134.09
Grade BAKER ANALYZED™ Biochemical Reagent
Quantity 500 g
Packaging Poly Bottle
Specific Gravity 1L = 1.60kg
Chemical Name or Material DL-Malic Acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (±)-Malic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: (±)-Malic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.