missing translation for 'onlineSavingsMsg'
Learn More

Dl-Malic Acid, Extra Pure, SLR, Fisher Chemical

Product Code. 10090240 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10090240 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10090240 Supplier Fisher Chemical Supplier No. M/1350/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 6915-15-7
Melting Point 128°C
Color White
pH 2
Packaging Plastic powder jar
Molecular Formula C4H6O5
MDL Number 64212
Quantity 500 g
Synonym malic acid, dl-malic acid, 2-hydroxysuccinic acid, malate, hydroxysuccinic acid, butanedioic acid, hydroxy, kyselina jablecna, pomalus acid, hydroxybutanedioic acid, deoxytetraric acid
InChI Key BJEPYKJPYRNKOW-UHFFFAOYSA-N
SMILES C(C(C(=O)O)O)C(=O)O
IUPAC Name 2-hydroxybutanedioic acid
Molecular Weight (g/mol) 134.087
PubChem CID 525
ChEBI CHEBI:6650
Formula Weight 134.09g/mol
Physical Form Solid
Show More Show Less
compliance-icons
Product Identifier
  • DL-Malic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.