missing translation for 'onlineSavingsMsg'
Learn More

DL-Mandelic acid, ≥99.0%, Honeywell Fluka™

Product Code. 15626090 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15626090 500 g Poly Bottle 500g
15616090 100 g Poly Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15626090 Supplier Honeywell-Fluka Supplier No. 63470500G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

for gravimetric determination of Zr,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-81-8
Molecular Formula Cl2MgO8
Molecular Weight (g/mol) 223.197
MDL Number MFCD00011102
InChI Key MPCRDALPQLDDFX-UHFFFAOYSA-L
Synonym anhydrone, magnesium perchlorate, dehydrite, perchloric acid, magnesium salt, anhydrous magnesium perchlorate, unii-7n77z541yf, hsdb 661, magnesium diperchlorate, magnesium perchlorate, anhydrous, perchlorate de magnesium french
PubChem CID 24840
IUPAC Name magnesium;diperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]

Specifications

Linear Formula Mg(ClO4)2
UN Number UN1475
Formula Weight 223.21g/mol
Percent Purity ≥99.0% (calc. based on dry substance, KT)
Quantity 500 g
Packaging Poly Bottle
Physical Form Solid
Chemical Name or Material Magnesium perchlorate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.