missing translation for 'onlineSavingsMsg'
Learn More

DL-Methotrexate-d3 Dipotassium Salt, TRC

Product Code. 30481743
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481743 10 mg 10mg
30484157 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30481743 Supplier Toronto Research Chemicals Supplier No. TRCM26068210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C20H17D3K2N8O5
Molecular Weight (g/mol) 533.64
Synonym N-[4-[[(2,4-Diamino-6-pteridinyl)methyl]methylamino]benzoyl]-glutamic Acid-d3 Dipotassium Salt, DL-Amethopterin-d3 Dipotassium Salt, NSC 117356-d3 Dipotassium Salt
SMILES O=C(NC(C(O[K])=O)CCC(O[K])=O)C(C=C1)=CC=C1N(C([2H])([2H])[2H])CC2=NC3=C(N)N=C(N)N=C3N=C2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H22N8O5.2K/c1-28(9-11-8-23-17-15(24-11)16(21)26-20(22)27-17)12-4-2-10(3-5-12)18(31)25-13(19(32)33)6-7-14(29)30;;/h2-5,8,13H,6-7,9H2,1H3,(H,25,31)(H,29,30)(H,32,33)(H4,21,22,23,26,27);;/q;2*+1/p-2/i1D3;;
Chemical Name or Material DL-Methotrexate-d3 Dipotassium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.